Yttrium (III) nitrate hydrate

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

ACTIAS23538G100EA 0 CHF
ACTIAS23538G100
Yttrium (III) nitrate hydrate
Yttrium (III) nitrate hydrate
Formula: N₃O₉Y·H₂O
MW: 274.92 g/mol (anhy)
MDL Number: MFCD00011469
CAS Number: 140191-40-8
EINECS: 233-802-6
UN: 1477
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR