Ytterbium (III) chloride hexahydrate

ACTIAS23558G100EA 0 CHF
ACTIAS23558G100
Ytterbium (III) chloride hexahydrate
Ytterbium (III) chloride hexahydrate
Formula: Cl₃Yb·6H₂O
MW: 387.49 g/mol
Melting Pt: 154 °C
Density: 2.57 g/cm³ (20 °C)
MDL Number: MFCD00149885
CAS Number: 10035-01-5
EINECS: 233-800-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR