1-Amino-2,2,2-trifluoroethane hydrochloride

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2,2,2-Trifluoroethylamine hydrochloride

ACTIAS24973G25EA 0 CHF
ACTIAS24973G25 ACTIAS24973G100
1-Amino-2,2,2-trifluoroethane hydrochloride
1-Amino-2,2,2-trifluoroethane hydrochloride
Formula: C₂H₅ClF₃N
MW: 135.52 g/mol
Melting Pt: 222…226 °C
MDL Number: MFCD00012875
CAS Number: 373-88-6
EINECS: 206-771-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR