(±)-3-Amino-1,2-propanediol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS20779G100EA 186 CHF
ACTIAS20779G100
(±)-3-Amino-1,2-propanediol
(±)-3-Amino-1,2-propanediol
Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 264 °C (1013 hPa)
Density: 1.1752 g/cm³ (20 °C)
MDL Number: MFCD00008140
CAS Number: 616-30-8
EINECS: 210-475-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR