3-Hydroxy-1-methylpiperidine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-Methylpiperidin-3-olN-Methyl-3-piperidinol

ACTIAS12610G25EA 0 CHF
ACTIAS12610G25
3-Hydroxy-1-methylpiperidine
3-Hydroxy-1-methylpiperidine
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 30…34 °C (0.1 mmHg)
Density: 0.9635 g/cm³ (16 °C)
MDL Number: MFCD00006495
CAS Number: 3554-74-3
EINECS: 222-609-2
https://ch.vwr.com/stibo/web/std.lang.all/53/47/2305347.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR