Ytterbium(III) oxide

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Diytterbium trioxideYtterbium oxide

ACTIAS20651G25EA 0 CHF
ACTIAS20651G25 ACTIAS20651G100
Ytterbium(III) oxide
Ytterbium (III) oxide
Formula: O₃Yb₂
MW: 394.08 g/mol
Melting Pt: 2346 °C
Density: 9.2 g/cm³ (20 °C)
MDL Number: MFCD00011290
CAS Number: 1314-37-0
EINECS: 215-234-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR