4-Methylcinnamic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3-(p-Tolyl)acrylic acid

ACTIAS21124G100EA 101 CHF
ACTIAS21124G100
4-Methylcinnamic acid
4-Methylcinnamic acid
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 303 °C (1013 hPa)
Melting Pt: 196…198 °C
MDL Number: MFCD00002697
CAS Number: 1866-39-3
EINECS: 217-479-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR