Samarium trichloride hexahydrate
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
Samarium trichloride hexahydrate
, Samarium(III) chloride hexahydrate
ACTIAS23556G100EA
0
CHF
ACTIAS23556G100
ACTIAS23556G25
Samarium trichloride hexahydrate
Samarium trichloride hexahydrate
Formula:
Cl₃Sm·6H₂O MW: 364.81 g/mol Density: 2.383 g/cm³ (25 °C) |
MDL Number:
MFCD00011235 CAS Number: 13465-55-9 EINECS: 233-797-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...