Hydrochloric acid 0.1 mol/l (0.1 N)

Danger

PRLSMC302590.0500EA 0 CHF
PRLSMC302590.0500 PRLSMC302590.1000
Hydrochloric acid 0.1 mol/l (0.1 N)
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 0.79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR