Indole-7-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1H-Indole-7-carboxylic acid

ACTIAS7103G5EA 68 CHF
ACTIAS7103G25 ACTIAS7103G1 ACTIAS7103G5
Indole-7-carboxylic acid
Indole-7-carboxylic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD00210442
CAS Number: 1670-83-3
EINECS: 216-801-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR