7-Chlorindol
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS40541G5EA
0
CHF
ACTIAS40541G5
7-Chlorindol
7-Chlorindol
Formel:
C₈H₆ClN MW: 151,6 g/mol |
MDL Number:
MFCD01321301 CAS-Nummer: 53924-05-3 EINECS: 258-865-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...