Indole-6-carboxylic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS7056G100EA
0
CHF
ACTIAS7056G25
ACTIAS7056G100
Indole-6-carboxylic acid
Indole-6-carboxylic acid
Formula:
C₉H₇NO₂ MW: 161.16 g/mol Melting Pt: 254…257 °C |
MDL Number:
MFCD00210441 CAS Number: 1670-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...