Indole-6-carbonitrile

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 6-Cyano-1H-indole6-Cyanoindole1H-Indole-6-carbonitrile

ACTIAS31578G10EA 0 CHF
ACTIAS31578G10 ACTIAS31578G5
Indole-6-carbonitrile
Indole-6-carbonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
MDL Number: MFCD00016732
CAS Number: 15861-36-6
EINECS: 239-988-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR