6-Chlorindol

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS7084G100EA 0 CHF
ACTIAS7084G5 ACTIAS7084G100
6-Chlorindol
6-Chlorindol
Formel: C₈H₆ClN
MW: 151,6 g/mol
Melting Pt: 90 °C
MDL Number: MFCD00005681
CAS-Nummer: 17422-33-2
EINECS: 241-449-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR