5-Methylindole

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS31955G5EA 0 CHF
ACTIAS31955G10 ACTIAS31955G5
5-Methylindole
5-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 60 °C
Density: 1,0202 g/cm³ (78.3 °C)
MDL Number: MFCD00005680
CAS Number: 614-96-0
EINECS: 210-400-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR