5-Brom-2-pentanon
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS34463G1EA
150
CHF
ACTIAS34463G1
ACTIAS34463G5
5-Brom-2-pentanon
5-Brom-2-pentanon
Formel:
H₃CC(O)(CH₂)₂CH₂Br MW: 165,03 g/mol |
MDL Number:
MFCD00051425 CAS-Nummer: 3884-71-7 EINECS: 223-419-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...