4-Phenylbutyric acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21488G100EA 0 CHF
ACTIAS21488G100
4-Phenylbutyric acid
4-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 290…294 °C (1013 hPa)
Melting Pt: 48…51 °C
MDL Number: MFCD00004403
CAS Number: 1821-12-1
EINECS: 217-341-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR