4-Methylindole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50151G5EA 0 CHF
ACTIAS50151G5 ACTIAS50151G25 ACTIAS50151G1
4-Methylindole
4-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 5 °C
Density: 1,062 g/cm³ (20 °C)
MDL Number: MFCD00005668
CAS Number: 16096-32-5
EINECS: 240-262-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR