4-Ethylbenzoyl chloride

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

ACTIAS24098G100EA 0 CHF
ACTIAS24098G100
4-Ethylbenzoyl chloride
4-Ethylbenzoyl chloride
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 235…236 °C (1013 hPa)
Melting Pt: 42…43 °C
Density: 1.14 g/cm³ (20 °C)
MDL Number: MFCD00000697
CAS Number: 16331-45-6
EINECS: 240-404-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR