4-Chlorophenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: (4-Chlorophenyl)metaboric acidDihydroxy(4-chlorophenyl)borane(4-Chlorophenyl)boronic acid4-Chlorobenzeneboronic acid

ACTIAS51136G5EA 0 CHF
ACTIAS51136G25 ACTIAS51136G5
4-Chlorophenylboronic acid
4-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 268…272 °C
Density: ∼1 g/cm³ (20 °C)
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR