4-Bromophenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: Dihydroxy(4-bromophenyl)borane(4-Bromophenyl)boronic acid(4-Bromophenyl)metaboric acid4-Bromobenzeneboronic acid

ACTIAS50032G100EA 0 CHF
ACTIAS50032G25 ACTIAS50032G100
4-Bromophenylboronic acid
4-Bromophenylboronic acid
Formula: C₆H₆BBrO₂
MW: 200.83 g/mol
Density: 1,67 g/cm³ (20 °C)
MDL Number: MFCD00002104
CAS Number: 5467-74-3
EINECS: 226-779-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR