4-Bromo-3-chloropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50422G5EA 0 CHF
ACTIAS50422G25 ACTIAS50422G5
4-Bromo-3-chloropyridine
4-Bromo-3-chloropyridine
Formula: C₅H₃BrClN
MDL Number: MFCD04112577
CAS Number: 73583-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR