4-Amino-2-bromopyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Bromopyridin-4-amine2-Bromo-4-pyridylamine

ACTIAS1009G5EA 0 CHF
ACTIAS1009G5
4-Amino-2-bromopyridine
4-Amino-2-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
MDL Number: MFCD01646061
CAS Number: 7598-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR