4,4,4-Trifluoro-1-butanol

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

ACTIAS23929G25EA 115 CHF
ACTIAS23929G25 ACTIAS23929G5
4,4,4-Trifluoro-1-butanol
4,4,4-Trifluoro-1-butanol
Formula: C₄H₇F₃O
MW: 128.09 g/mol
Boiling Pt: 122 °C (1013 hPa)
Density: 1.217 g/cm³ (25 °C)
MDL Number: MFCD00041483
CAS Number: 461-18-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR