5-Fluoro-m-toluic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS32395G10EA 0 CHF
ACTIAS32395G10 ACTIAS32395G1 ACTIAS32395G5
5-Fluoro-m-toluic acid
5-Fluoro-m-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
MDL Number: MFCD03094313
CAS Number: 518070-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR