3-Chlor-p-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

ACTIAS1495G5EA 0 CHF
ACTIAS1495G25 ACTIAS1495G5
3-Chlor-p-toluylsäure
3-Chlor-p-toluylsäure
Formel: C₈H₇ClO₂
MW: 170,6 g/mol
MDL Number: MFCD00045841
CAS-Nummer: 5162-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR