3-Bromphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

ACTIAS50031G5EA 0 CHF
ACTIAS50031G5 ACTIAS50031G25 ACTIAS50031G1
3-Bromphenylboronsäure
3-Bromphenylboronsäure
Formel: C₆H₆BBrO₂
MW: 200,83 g/mol
MDL Number: MFCD00239386
CAS-Nummer: 89598-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR