5-Bromo-2,3-dichloropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS7140G5EA 0 CHF
ACTIAS7140G25 ACTIAS7140G5
5-Bromo-2,3-dichloropyridine
5-Bromo-2,3-dichloropyridine
Formula: Câ‚…Hâ‚‚BrClâ‚‚N
MDL Number: MFCD04112492
CAS Number: 97966-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR