3-Amino-5-bromopyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 5-Bromopyridin-3-amine5-Bromo-3-pyridylamine

ACTIAS51660G1EA 0 CHF
ACTIAS51660G5 ACTIAS51660G1
3-Amino-5-bromopyridine
3-Amino-5-bromopyridine
Formula: Câ‚…Hâ‚…BrNâ‚‚
MW: 173.01 g/mol
MDL Number: MFCD01646060
CAS Number: 13535-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR