3-Amino-2-chlorpyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS31914G100EA 0 CHF
ACTIAS31914G100
3-Amino-2-chlorpyridin
3-Amino-2-chlorpyridin
Formel: C₅H₅ClN₂
MW: 128,56 g/mol
Siedepunkt: 263 °C (1013 hPa)
Melting Pt: 78…79 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00006238
CAS-Nummer: 6298-19-7
EINECS: 228-572-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR