3,5-Difluorphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

ACTIAS50265G100EA 0 CHF
ACTIAS50265G100 ACTIAS50265G5
3,5-Difluorphenylboronsäure
3,5-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
MW: 157,91 g/mol
MDL Number: MFCD01318138
CAS-Nummer: 156545-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR