Pyridine-3,4-dicarboxylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 3,4-Pyridinedicarboxylic acidCinchomeronic acidPyridine-3,4-dicarboxylicacid

ACTIAS4018G100EA 0 CHF
ACTIAS4018KG1 ACTIAS4018G100
Pyridine-3,4-dicarboxylic acid
Pyridine-3,4-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 529 °C (1013 hPa)
Melting Pt: 257 °C
Storage Temperature: Ambient
MDL Number: MFCD00006392
CAS Number: 490-11-9
EINECS: 207-705-4
Merck Index: 13,02304

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR