2-Fluoro-6-methylpyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: 2-Fluoro-6-picoline

ACTIAS40243G100EA 0 CHF
ACTIAS40243G100
2-Fluoro-6-methylpyridine
2-Fluoro-6-methylpyridine
Formula: C₆H₆FN
MW: 111.12 g/mol
MDL Number: MFCD00041226
CAS Number: 407-22-7
EINECS: 206-980-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR