2-Bromopyrimidine
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Warning
|
ACTIAS50196G5EA
0
CHF
ACTIAS50196G5
ACTIAS50196G100
2-Bromopyrimidine
2-Bromopyrimidine
Formula:
C₄H₃BrN₂ MW: 158.99 g/mol Boiling Pt: 62…64 °C (1,5 mmHg) Melting Pt: 56…57,5 °C Density: >1 g/cm³ (20 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00014601 CAS Number: 4595-60-2 EINECS: 224-993-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...