2-Bromo-5-fluoropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS4050G5EA 0 CHF
ACTIAS4050G5 ACTIAS4050G25
2-Bromo-5-fluoropyridine
2-Bromo-5-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
MDL Number: MFCD00234011
CAS Number: 41404-58-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR