2-Bromo-1,1-diethoxypropane

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS51640G5EA 152 CHF
ACTIAS51640G5 ACTIAS51640G100 ACTIAS51640G25
2-Bromo-1,1-diethoxypropane
2-Bromo-1,1-diethoxypropane
Formula: (H₃CCH₂O)₂HCBrCHCH₃
MDL Number: MFCD00152299
CAS Number: 3400-55-3
EINECS: 222-270-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR