2-Amino-6-fluorpyridin
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS40164G5EA
0
CHF
ACTIAS40164G5
ACTIAS40164G100
ACTIAS40164G25
2-Amino-6-fluorpyridin
2-Amino-6-fluorpyridin
Formel:
C₅H₅FN₂ MW: 112,11 g/mol |
MDL Number:
MFCD04114200 CAS-Nummer: 1597-32-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...