2-Amino-5-iodopyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 5-Iodo-2-pyridylamine5-Iodopyridin-2-amine

ACTIAS4027G100EA 0 CHF
ACTIAS4027G100 ACTIAS4027G5
2-Amino-5-iodopyridine
2-Amino-5-iodopyridine
Formula: Câ‚…Hâ‚…INâ‚‚
MW: 220.01 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 128…131 °C
MDL Number: MFCD00160312
CAS Number: 20511-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR