2-Amino-5-brom-4-methylpyridin
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS50989G5EA
0
CHF
ACTIAS50989G1
ACTIAS50989G5
ACTIAS50989G25
2-Amino-5-brom-4-methylpyridin
2-Amino-5-brom-4-methylpyridin
Formel:
C₆H₇BrN₂ MW: 187,04 g/mol |
MDL Number:
MFCD03427660 CAS-Nummer: 98198-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...