Pyridine-2,6-dicarboxylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 2,6-Pyridinedicarboxylic acidDipicolinic acidDPA

ACTIAS7023G100EA 0 CHF
ACTIAS7023G100 ACTIAS7023KG1
Pyridine-2,6-dicarboxylic acid
Pyridine-2,6-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 327 °C
Flash Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006299
CAS Number: 499-83-2
EINECS: 207-894-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR