2,6-Dimethylphenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Dihydroxy(2,6-dimethylphenyl)borane(2,6-Dimethylphenyl)metaboric acid(2,6-Dimethylphenyl)boronic acid2,6-Dimethylbenzeneboronic acid

ACTIAS32217G1EA 0 CHF
ACTIAS32217G10 ACTIAS32217G1
2,6-Dimethylphenylboronic acid
2,6-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD01009693
CAS Number: 100379-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR