2,5-Difluorophenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50245G5EA 0 CHF
ACTIAS50245G5 ACTIAS50245G25
2,5-Difluorophenylboronic acid
2,5-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
MDL Number: MFCD01863171
CAS Number: 193353-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR