2,4-Dichloropyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS7004G100EA 0 CHF
ACTIAS7004KG1 ACTIAS7004G100
2,4-Dichloropyrimidine
2,4-Dichloropyrimidine
Formula: Câ‚„Hâ‚‚Clâ‚‚Nâ‚‚
MW: 148.98 g/mol
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: 61 °C
MDL Number: MFCD00006061
CAS Number: 3934-20-1
EINECS: 223-508-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR