2,3-Difluorpyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH
Gefahr

ACTIAS51300G5EA 0 CHF
ACTIAS51300G5 ACTIAS51300G1 ACTIAS51300G25
2,3-Difluorpyridin
2,3-Difluoropyridine
Formel: C₅H₃F₂N
MW: 115,08 g/mol
MDL Number: MFCD04114143
CAS-Nummer: 1513-66-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR