2,2-Difluoroethanol

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21389G25EA 160 CHF
ACTIAS21389G100 ACTIAS21389G25
2,2-Difluoroethanol
2,2-Difluoroethanol
Formula: Câ‚‚Hâ‚„Fâ‚‚O
MW: 82.05 g/mol
Boiling Pt: 95 °C (1013 hPa)
Melting Pt: –28.2 °C
Density: 1.296 g/cm³ (20 °C)
MDL Number: MFCD00042248
CAS Number: 359-13-7
UN: 1987
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR