11-Bromo-1-undecanol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 11-Bromoundecanol11-Bromoundecan-1-ol

ACTIAS23512G100EA 161 CHF
ACTIAS23512G100 ACTIAS23512G25
11-Bromo-1-undecanol
11-Bromo-1-undecanol
Formula: HO(CH₂)₁₀CH₂Br
MW: 251.21 g/mol
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00004752
CAS Number: 1611-56-9
EINECS: 216-554-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR