4-(Trifluormethyl)anisol
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS34985G5EA
0
CHF
ACTIAS34985G1
ACTIAS34985G5
4-(Trifluormethyl)anisol
4-(Trifluormethyl)anisol
Formel:
C₈H₇F₃O MW: 176,14 g/mol |
MDL Number:
MFCD03094456 CAS-Nummer: 402-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...