1,1,2,2-Tetrafluoro-1-methoxyethane

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS24896G100EA 0 CHF
ACTIAS24896G100 ACTIAS24896G25
1,1,2,2-Tetrafluoro-1-methoxyethane
1,1,2,2-Tetrafluoro-1-methoxyethane
Formula: C₃H₄F₄O
MW: 132.06 g/mol
Boiling Pt: 33 °C (1013 hPa)
Melting Pt: –107 °C
Density: 1.2939 g/cm³ (20 °C)
MDL Number: MFCD00042106
CAS Number: 425-88-7
EINECS: 207-039-4
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR