2'-Fluoroacetophenone

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: o-Fluoroacetophenone1-(2-Fluorophenyl)ethanone

ACTIAS23326G100EA 146 CHF
ACTIAS23326G100
2'-Fluoroacetophenone
2'-Fluoroacetophenone
Formula: C₈H₇FO
MW: 138.14 g/mol
Boiling Pt: 82…83 °C (13,3 hPa)
Density: 1,12 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000320
CAS Number: 445-27-2
EINECS: 207-156-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR