1-Chloro-4-methoxybutane

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS24465G100EA 105 CHF
ACTIAS24465G100 ACTIAS24465G25
1-Chloro-4-methoxybutane
1-Chloro-4-methoxybutane
Formula: C₅H₁₁ClO
MW: 122.59 g/mol
MDL Number: MFCD00209961
CAS Number: 17913-18-7
EINECS: 241-858-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR